2,4-Dimethyl-5-(1H-pyrazol-3-yl)-1,3-thiazole

Supplier: APOLLO SCIENTIFIC
Warning

OR110950-1G OR110950-5G
77403-584EA 176.26 USD
77403-584 77403-586
2,4-Dimethyl-5-(1H-pyrazol-3-yl)-1,3-thiazole
2,4-Dimethyl-5-(1H-pyrazol-3-yl)-1,3-thiazole
Formula: C₈H₉N₃S
MW: 179.242 g/mol
Melting Pt: 161…164 °C
Storage Temperature: Ambient
MDL Number: MFCD01763614
CAS Number: 439107-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR