Methyl 3-(4,5-dimethoxy-2-nitrophenyl)acrylate

Supplier: APOLLO SCIENTIFIC

OR110892-10G OR110892-1G OR110892-5G
77403-556EA 550.16 USD
77403-556 77403-558 77403-560
Methyl 3-(4,5-dimethoxy-2-nitrophenyl)acrylate
Methyl 3-(4,5-dimethoxy-2-nitrophenyl)acrylate
Formula: C₁₂H₁₃NO₆
MW: 267.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00266189
CAS Number: 118509-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR