4-(4-Methylphenoxy)-1H-indazol-3-amine

Supplier: APOLLO SCIENTIFIC

OR110880-1G OR110880-250MG OR110880-5G
77403-538EA 330.1 USD
77403-538 77403-540 77403-542
4-(4-Methylphenoxy)-1H-indazol-3-amine
4-(4-Methylphenoxy)-1H-indazol-3-amine
Formula: C₁₄H₁₃N₃O
Storage Temperature: Ambient
MDL Number: MFCD26939170
CAS Number: 871708-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR