5-Bromo-N-methoxy-N-methyl-2-furamide

Supplier: APOLLO SCIENTIFIC
Warning

OR110852-1G OR110852-500MG
77403-516EA 340.76 USD
77403-516 77403-518
5-Bromo-N-methoxy-N-methyl-2-furamide
5-Bromo-N-methoxy-N-methyl-2-furamide
Formula: C₇H₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD16104416
CAS Number: 179055-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR