4-(4-Bromophenyl)-1-isopropyl-5-oxo-4,5-dihydro-1H-1,2,4-triazole-3-carboxylic acid

Supplier: APOLLO SCIENTIFIC

OR110811-1G OR110811-250MG
77403-488EA 406.5 USD
77403-488 77403-490
4-(4-Bromophenyl)-1-isopropyl-5-oxo-4,5-dihydro-1H-1,2,4-triazole-3-carboxylic acid
4-(4-Bromophenyl)-1-isopropyl-5-oxo-4,5-dihydro-1H-1,2,4-triazole-3-carboxylic acid
Formula: C₁₂H₁₂BrN₃O₃
MW: 326.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18074539
CAS Number: 1427460-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR