2-Amino-5-methoxyphenol

Supplier: APOLLO SCIENTIFIC
Warning

OR110697-1G OR110697-250MG
77403-418EA 128.38 USD
77403-418 77403-420
2-Amino-5-methoxyphenol
2-Amino-5-methoxyphenol
Formula: C₇H₉NO₂
Melting Pt: 126…129 °C
Storage Temperature: Freezer
MDL Number: MFCD03840435
CAS Number: 40925-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR