N-Methyl-4-pentylbenzamide

Supplier: APOLLO SCIENTIFIC

OR110452-5G OR110452-1G OR110452-250MG
77403-354EA 705.02 USD
77403-354 77403-350 77403-352
N-Methyl-4-pentylbenzamide
N-Methyl-4-pentylbenzamide
Formula: C₁₃H₁₉NO
MW: 205.296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03551083
CAS Number: 401587-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR