4-Pentylbenzamide

Supplier: APOLLO SCIENTIFIC

OR110451-1G OR110451-250MG OR110451-5G
77403-344EA 176.26 USD
77403-344 77403-346 77403-348
4-Pentylbenzamide
4-Pentylbenzamide
Formula: C₁₂H₁₇NO
Storage Temperature: Ambient
MDL Number: MFCD00984217
CAS Number: 138913-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR