4-(3-Bromophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one

Supplier: APOLLO SCIENTIFIC
Warning

OR110421-10G OR110421-1G OR110421-5G
77403-324EA 686.68 USD
77403-324 77403-326 77403-328
4-(3-Bromophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
4-(3-Bromophenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Formula: C₈H₆BrN₃O
MW: 240.057 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384918
CAS Number: 1007578-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR