3-Bromo-1H-indazol-7-amine
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR110360-1G
OR110360-250MG
77403-282EA
212.94
USD
77403-282
77403-284
3-Bromo-1H-indazol-7-amine
3-Bromo-1H-indazol-7-amine
Formula:
C₇H₆BrN₃ MW: 212.047 g/mol Melting Pt: 135 °C Storage Temperature: Ambient |
MDL Number:
MFCD09027010 CAS Number: 316810-90-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...