5-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,3-oxazole

Supplier: APOLLO SCIENTIFIC

OR110279-1G OR110279-250MG
77403-222EA 275.08 USD
77403-222 77403-224
5-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,3-oxazole
5-(2,3-Dihydro-1,4-benzodioxin-6-yl)-1,3-oxazole
Formula: C₁₁H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD13191320
CAS Number: 1305712-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR