Methyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR110272-1G OR110272-5G
77403-208EA 195.06 USD
77403-208 77403-210
Methyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate
Methyl 3-amino-4-methoxy-1-benzothiophene-2-carboxylate
Formula: C₁₁H₁₁NO₃S
Storage Temperature: Ambient
MDL Number: MFCD08706234
CAS Number: 35212-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR