Methyl 6-amino-5-chloronicotinate

Supplier: APOLLO SCIENTIFIC
Danger

OR110263-5G OR110263-1G
77403-194EA 447.26 USD
77403-194 77403-192
Methyl 6-amino-5-chloronicotinate
Methyl 6-amino-5-chloronicotinate
Formula: C₇H₇ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12964045
CAS Number: 856211-63-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR