Ethyl 2,5-dimethyl-1H-pyrrole-3-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR110260-1G OR110260-5G
77403-188EA 66.22 USD
77403-188 77403-190
Ethyl 2,5-dimethyl-1H-pyrrole-3-carboxylate
Ethyl 2,5-dimethyl-1H-pyrrole-3-carboxylate
Formula: C₉H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00022376
CAS Number: 2199-52-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR