Methyl 2-chloro-6-(piperazin-1-yl)isonicotinate

Supplier: APOLLO SCIENTIFIC
Warning

OR110254-1G OR110254-250MG
77403-182EA 303.6 USD
77403-182 77403-184
Methyl 2-chloro-6-(piperazin-1-yl)isonicotinate
Methyl 2-chloro-6-(piperazin-1-yl)isonicotinate
Formula: C₁₁H₁₄ClN₃O₂
MW: 255.701 g/mol
Melting Pt: 72…74 °C
Storage Temperature: Ambient
MDL Number: MFCD12545990
CAS Number: 1227954-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR