4-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one

Supplier: APOLLO SCIENTIFIC

OR110244-1G OR110244-250MG
77403-172EA 209.88 USD
77403-172 77403-174
4-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one
4-Methyl-5-nitro-2H-1,4-benzoxazin-3(4H)-one
Formula: C₉H₈N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD12406815
CAS Number: 132522-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR