N,N-Diethyl-1,3,4-thiadiazole-2,5-diamine

Supplier: APOLLO SCIENTIFIC

OR110239-1G OR110239-250MG
77403-164EA 330.1 USD
77403-164 77403-166
N,N-Diethyl-1,3,4-thiadiazole-2,5-diamine
N,N-Diethyl-1,3,4-thiadiazole-2,5-diamine
Formula: C₆H₁₂N₄S
MW: 172.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756249
CAS Number: 894880-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR