2-Chloro-1-ethyl-1H-indole-3-carbaldehyde

Supplier: APOLLO SCIENTIFIC

OR110223-250MG OR110223-1G
77403-154EA 75.4 USD
77403-154 77403-152
2-Chloro-1-ethyl-1H-indole-3-carbaldehyde
2-Chloro-1-ethyl-1H-indole-3-carbaldehyde
Formula: C₁₁H₁₀ClNO
MW: 207.656 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02253188
CAS Number: 64788-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR