2-Methyl-6-nitro-2H-indazole

Supplier: APOLLO SCIENTIFIC

OR110174-1G OR110174-5G
77403-102EA 146.7 USD
77403-102 77403-104
2-Methyl-6-nitro-2H-indazole
2-Methyl-6-nitro-2H-indazole
Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00456075
CAS Number: 6850-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR