4-(1-Methyl-1H-indazol-3-yl)-2,4-dihydro-3H-1,2,4-triazol-3-one

Supplier: APOLLO SCIENTIFIC

OR110107-1G OR110107-250MG
77403-078EA 330.1 USD
77403-078 77403-080
4-(1-Methyl-1H-indazol-3-yl)-2,4-dihydro-3H-1,2,4-triazol-3-one
4-(1-Methyl-1H-indazol-3-yl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Formula: C₁₀H₉N₅O
MW: 215.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23381088
CAS Number: 1440535-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR