Dimethyl [2-(methoxycarbonyl)ethyl]phosphonate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Trimethyl 3-phosphonopropionate, Methyl 3-(dimethoxyphosphoryl)propanoate

OR10767-1G OR10767-5G
77402-914EA 54.5 USD
77402-914 77402-916
Dimethyl [2-(methoxycarbonyl)ethyl]phosphonate
Dimethyl [2-(methoxycarbonyl)ethyl]phosphonate
Formula: C₆H₁₃O₅P
MW: 196.138 g/mol
Boiling Pt: 137…140 °C (10 mm Hg)
Density: 1.21 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00137948
CAS Number: 18733-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR