Diethyl (3-chloro-2-oxoprop-1-yl)phosphonate

Supplier: APOLLO SCIENTIFIC
Warning

OR10698-1G OR10698-250MG
77402-896EA 922.46 USD
77402-896 77402-898
Diethyl (3-chloro-2-oxoprop-1-yl)phosphonate
Diethyl (3-chloro-2-oxoprop-1-yl)phosphonate
Formula: C₇H₁₄ClO₄P
Boiling Pt: 94…95 °C (0.07 mm Hg)
Density: 1.219 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD04038926
CAS Number: 67257-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR