N-Benzyl-2-(piperazin-1-yl)acetamide dihydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Piperazino-acetic acid-benzylamide dihydrochloride

OR0245-250MG OR0245-500MG
77401-966EA 21.26 USD
77401-966 77401-968
N-Benzyl-2-(piperazin-1-yl)acetamide dihydrochloride 97%
N-Benzyl-2-(piperazin-1-yl)acetamide dihydrochloride
Formula: C₁₃H₁₉N₃O·2ClH
Melting Pt: 218…221 °C
Storage Temperature: Ambient
MDL Number: MFCD04115628
CAS Number: 827614-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR