4-[(Piperazin-1-yl)methyl]benzonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR0230-1G OR0230-5G
77401-950EA 117.16 USD
77401-950 77401-952
4-[(Piperazin-1-yl)methyl]benzonitrile 97%
4-[(Piperazin-1-yl)methyl]benzonitrile
Formula: C₁₂H₁₅N₃
Storage Temperature: Ambient
MDL Number: MFCD04972584
CAS Number: 89292-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR