3-(4-Chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Carboxy-4-(4-chlorophenyl)-2-(methysulphanyl)thiophene-3-carbonitrile, 3-(4-Chlorophenyl)-4-cyano-5-(methylthio)-2-thenoic acid

OR018945-1G OR018945-250MG
77401-854EA 137.54 USD
77401-854 77401-856
3-(4-Chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid 97%
3-(4-Chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid
Formula: C₁₃H₈ClNO₂S₂
MW: 309.791 g/mol
Melting Pt: 241…243 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00112478
CAS Number: 116525-66-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR