4-Hydroxybenzimidazole 95%

Supplier: APOLLO SCIENTIFIC

OR943594-1G OR943594-250MG
77412-108EA 85.58 USD
77412-108 77412-110
4-Hydroxybenzimidazole 95%
4-Hydroxybenzimidazole
Formula: C₇H₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD00963978
CAS Number: 67021-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR