Methyl 4-Bromo-2-chlorobenzoate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Methyl 4-bromo-2-chlorobenzoate, 4-Bromo-2-chlorobenzoic acid methyl ester

OR17252-10G OR17252-1G OR17252-5G
77405-988EA 93.74 USD
77405-988 77405-990 77405-992
Methyl 4-Bromo-2-chlorobenzoate 97%
Methyl 4-Bromo-2-chlorobenzoate
Formula: C₈H₆BrClO₂
MW: 249.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566823
CAS Number: 185312-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR