Pentafluorophenyl Benzoate 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC908537-1G PC908537-5G
77418-542EA 33.62 USD
77418-542 77418-544
Pentafluorophenyl Benzoate 98%
Pentafluorophenyl Benzoate
Formula: C₁₃H₅F₅O₂
Melting Pt: 73…74 °C
Storage Temperature: Refrigerator
CAS Number: 1548-84-1
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR