3-Chloro-N,N-diethylpropan-1-amine
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR315510-25G
OR315510-5G
77409-088EA
81.5
USD
77409-088
77409-090
3-Chloro-N,N-diethylpropan-1-amine
3-Chloro-N,N-diethylpropan-1-amine
Formula:
C₇H₁₆ClN Boiling Pt: 169…171 °C/760 mmHg Density: 0.89 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00039385 CAS Number: 104-77-8 UN: 1992 ADR: 3+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...