H-4-Pal-OH·2HCl 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-(4-Pyridyl)-alanine·2HCl, H-4-Pal-OH, L-4-Pyridylalanine

OR14725-1G OR14725-250MG OR14725-5G
77405-100EA 124.3 USD
77405-100 77405-102 77405-104
H-4-Pal-OH·2HCl 98%
H-4-Pal-OH·2HCl
Formula: C₈H₁₀N₂O₂·₂HCl
MW: 239.2 g/mol
Melting Pt: 257…259 °C
Storage Temperature: Ambient
MDL Number: MFCD01860883
CAS Number: 37535-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR