Bis(trifluoromethanesulfonyl)imide 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
lithium bis(trifluoromethanesulphonimide), 1,1,1-Trifluoro-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N,N-Bis(trifluoromethylsulphonyl)amine
PC0574-100G
PC0574-25G
77412-636EA
447.26
USD
77412-636
77412-638
Bis(trifluoromethanesulfonyl)imide 98%
Bis(trifluoromethanesulfonyl)imide
Formula:
C₂HF₆NO₄S₂ MW: 281.143 g/mol Boiling Pt: 90…91 °C Melting Pt: 46…57 °C Density: 1.36 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00214154 CAS Number: 82113-65-3 UN: 3261 ADR: 8,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...