2-Fluoro-5-methoxy-3-(trifluoromethyl)aniline 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-fluoro-5-methoxy-3-(trifluoromethyl)aniline

PC303034-1G PC303034-5G
77414-344EA 83.54 USD
77414-344 77414-346
2-Fluoro-5-methoxy-3-(trifluoromethyl)aniline 97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)aniline
Formula: C₈H₇F₄NO
MW: 209.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201043
CAS Number: 1262198-05-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR