2-Fluoro-5-methyl-4-(trifluoromethyl)phenylacetonitrile 98%

Supplier: APOLLO SCIENTIFIC

Synonyms: 2-Fluoro-5-methyl-4-(trifluoromethyl)phenylacetonitrile

PC302808-5G PC302808-1G
77414-274EA 895.54 USD
77414-274 77414-272
2-Fluoro-5-methyl-4-(trifluoromethyl)phenylacetonitrile 98%
2-Fluoro-5-methyl-4-(trifluoromethyl)phenylacetonitrile
Formula: C₁₀H₇F₄N
MW: 217.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687199
CAS Number: 1323966-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR