N-Methylpyrrole-2-boronic acid pinacol ester 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole, N-METHYLPYRROLE-2-BORONIC ACID, PINACOL ESTER, 1-Methyl-1H-pyrrole-2-boronic acid, pinacol ester

OR1189-500MG OR1189-5G OR1189-1G
77404-054EA 34.64 USD
77404-054 77404-056 77404-052
N-Methylpyrrole-2-boronic acid pinacol ester 97%
N-Methylpyrrole-2-boronic acid pinacol ester
Formula: C₁₁H₁₈BNO₂
MW: 207.08 g/mol
Melting Pt: 44…50 °C
Flash Pt: 82 °C
Storage Temperature: Freezer
MDL Number: MFCD06659917
CAS Number: 850567-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR