2-(2-Propynyloxy)tetrahydro-2H-pyran 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Tetrahydro-2-(2-propynyloxy)-2H-pyran, 2-Propargyloxane, 2-(Prop-2-yn-1-yloxy)tetrahydro-2H-pyran, Propargyl 2-tetrahydropyranyl ether, 2-(2-Propynyloxy)tetrahydropyran

OR471618-100G OR471618-25G OR471618-5G
77409-974EA 124.3 USD
77409-974 77409-976 77409-978
2-(2-Propynyloxy)tetrahydro-2H-pyran 99%
2-(2-Propynyloxy)tetrahydro-2H-pyran
Formula: C₈H₁₂O₂
MW: 140.18 g/mol
Boiling Pt: 65 °C
Density: 1.01 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00006604
CAS Number: 6089-04-9
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR