N-Phenylhydroxylamine 95%

Supplier: APOLLO SCIENTIFIC

OR933342-1G OR933342-250MG OR933342-5G
77411-996EA 234.32 USD
77411-996 77411-998 77412-000
N-Phenylhydroxylamine 95%
N-Phenylhydroxylamine
Formula: C₆H₇NO
MW: 109.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045718
CAS Number: 100-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR