Tris[4-(trifluoromethyl)phenyl]phosphine 95+%

Supplier: APOLLO SCIENTIFIC

PC7919G-10G PC7919G-1G PC7919G-5G
77418-296EA 309.72 USD
77418-296 77418-298 77418-300
Tris[4-(trifluoromethyl)phenyl]phosphine 95+%
Tris[4-(trifluoromethyl)phenyl]phosphine
Formula: C₂₁H₁₂F₉P
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00058883
CAS Number: 13406-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR