Ethyl-4-(1-piperazinyl)benzoate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR0944-100G OR0944-25G OR0944-5G
77402-324EA 406.5 USD
77402-324 77402-326 77402-328
Ethyl-4-(1-piperazinyl)benzoate 98%
Ethyl-4-(1-piperazinyl)benzoate
Formula: C₁₃H₁₈N₂O₂
Melting Pt: 89…90 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04973340
CAS Number: 80518-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR