Ethyl-3-amino-4,4,4-trifluorobutyrate 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC4132-1G PC4132-5G
77415-156EA 108.28 USD
77415-156 77415-158 77415-160
Ethyl-3-amino-4,4,4-trifluorobutyrate 98%
Ethyl-3-amino-4,4,4-trifluorobutyrate
Formula: C₆H₁₀F₃NO₂
MW: 185.15 g/mol
Boiling Pt: 70…71 °C/4 mmHg
Storage Temperature: Ambient
MDL Number: MFCD01075754
CAS Number: 170804-18-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR