4-Chloro-3-(trifluoromethylthio)benzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC302573-25G PC302573-50G PC302573-5G
77414-198EA 425.86 USD
77414-198 77414-200 77414-202
4-Chloro-3-(trifluoromethylthio)benzyl bromide 97%
4-Chloro-3-(trifluoromethylthio)benzyl bromide
Formula: C₈H₅BrClF₃S
MW: 305.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660314
CAS Number: 886503-38-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR