4,5-Dimethoxy-o-phenylenediamine dihydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4,5-Dimethoxybenzene-1,2-diamine dihydrochloride

OR17128-1G OR17128-250MG
77405-968EA 200.7 USD
77405-968 77405-970
4,5-Dimethoxy-o-phenylenediamine dihydrochloride
4,5-Dimethoxy-o-phenylenediamine dihydrochloride
Formula: C₈H₁₂N₂O₂·2HCl
MW: 241.12 g/mol
Melting Pt: 266…268 °C
Storage Temperature: Ambient
MDL Number: MFCD00190674
CAS Number: 131076-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR