4-(Pentafluorothio)benzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(Bromomethyl)phenylsulphur pentafluoride

PC405656-500MG PC405656-5G PC405656-1G
77415-074EA 268.96 USD
77415-074 77415-076 77415-072
4-(Pentafluorothio)benzyl bromide
4-(Pentafluorothio)benzyl bromide
Formula: C₇H₆BrF₅S
MW: 297.08 g/mol
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD16652420
CAS Number: 1126969-29-6
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR