3-(Trifluoromethyl)crotonic acid 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC7668-25G PC7668-5G PC7668-100MG
77418-094EA 461.52 USD
77418-094 77418-096 77418-092
3-(Trifluoromethyl)crotonic acid 95%
3-(Trifluoromethyl)crotonic acid
Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
Boiling Pt: 158…163 °C
Melting Pt: 28…30 °C
Density: 1.318 g/cm³ (25 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00014374
CAS Number: 69056-67-3
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR