2-Chloro-1-(ethoxymethyl)imidazole

Supplier: APOLLO SCIENTIFIC
Warning

OR10410-100MG OR10410-1G OR10410-5G
77402-700EA 48.9 USD
77402-700 77402-702 77402-704
2-Chloro-1-(ethoxymethyl)imidazole
2-Chloro-1-(ethoxymethyl)imidazole
Formula: C₆H₉ClN₂O
MW: 160.60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788569
CAS Number: 850429-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR