2-Bromo-6-fluorophenylacetonitrile

Supplier: APOLLO SCIENTIFIC
Danger

PC408235-1G PC408235-250MG
77415-108EA 221.08 USD
77415-108 77415-110
2-Bromo-6-fluorophenylacetonitrile
2-Bromo-6-fluorophenylacetonitrile
Formula: C₈H₅BrFN
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD11226884
CAS Number: 936693-22-0
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR