1-Boc-azetidine-3-carboxaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Formylazetidine-1-carboxylic acid tert-Butyl ester, 1-(tert-Butoxycarbonyl)azetidine-3-carboxaldehyde, tert-Butyl 3-Formylazetidine-1-carboxylate, 1-Boc-azetidine-3-carboxaldehyde

OR15687-1G OR15687-25G OR15687-5G
77405-538EA 27.5 USD
77405-538 77405-540 77405-542
1-Boc-azetidine-3-carboxaldehyde
1-Boc-azetidine-3-carboxaldehyde
Formula: C₉H₁₅NO₃
Boiling Pt: 77…82 °C/2 mmHg
Storage Temperature: Freezer
MDL Number: MFCD06656142
CAS Number: 177947-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR