4-Methyl-1,3-thiazole-5-sulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-Methylthiazole-5-sulphonyl chloride

OR110665-100MG OR110665-1G OR110665-250MG
77403-404EA 38.72 USD
77403-404 77403-406 77403-408
4-Methyl-1,3-thiazole-5-sulfonyl chloride
4-Methyl-1,3-thiazole-5-sulfonyl chloride
Formula: C₄H₄ClNO₂S₂
Melting Pt: 34…36 °C
Storage Temperature: Freezer
MDL Number: MFCD09864968
CAS Number: 953070-51-4
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR