6-Bromo-7-methylquinoxaline

Supplier: APOLLO SCIENTIFIC
Warning

OR111498-1G OR111498-250MG
77403-788EA 894.52 USD
77403-788 77403-790
6-Bromo-7-methylquinoxaline
6-Bromo-7-methylquinoxaline
Formula: C₉H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD16987756
CAS Number: 646504-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR