2,6-Difluoro-4-methoxybenzoyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC8313-10G PC8313-1G PC8313-5G
77418-382EA 188.48 USD
77418-382 77418-384 77418-386
2,6-Difluoro-4-methoxybenzoyl chloride 97%
2,6-Difluoro-4-methoxybenzoyl chloride
Formula: C₈H₅ClF₂O₂
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD03094495
CAS Number: 125369-56-4
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR