Azetidin-2-one 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Azetidinone

OR24812-1G OR24812-5G
77407-538EA 65.2 USD
77407-538 77407-540
Azetidin-2-one 98%
Azetidin-2-one
Formula: C₃H₅NO
MW: 71.08 g/mol
Boiling Pt: 106 °C/15 mmHg
Melting Pt: 72…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013328
CAS Number: 930-21-2
UN: 3263
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR